accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10504-04-8

CAS RN 10504-04-8

IUPAC Name: 2,3,5-trimethylfuran
Synonyms: 2,3,5-trimethylfuran
Molecular Weight (g/mol): 110.16
Molecular Formula: C7H10O
InChi Key: NJXZFRUNHWKHEC-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C)O1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products