man

Test

CAS RN 10510-54-0

CH3OOH2NNNH2O

CAS RN 10510-54-0

IUPAC Name: 8,12-diamino-10λ⁴-oxa-5-azatetraphen-10-ylium acetate
Synonyms: cresyl violet acetate
Molecular Weight (g/mol): 321.34
Molecular Formula: C18H15N3O3
InChi Key: XKOCOMKJPWEOHX-UHFFFAOYSA-M
SMILES: CC([O-])=O.NC1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C(N)=C3
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Color 
  • (6)
  • (1)
Grade 
  • (5)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (1)
  • (5)