man

Test

CAS RN 105184-38-1

OOFF

CAS RN 105184-38-1

IUPAC Name: 2-(3,5-difluorophenyl)acetate
Synonyms: 2-(3,5-difluorophenyl)acetate
Molecular Weight (g/mol): 171.12
Molecular Formula: C8H5F2O2
InChi Key: IGGNSAVLXJKCNH-UHFFFAOYSA-M
SMILES: [O-]C(=O)CC1=CC(F)=CC(F)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)