accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10531-80-3

CAS RN 10531-80-3

IUPAC Name: 6-methyl-1,3-benzoxazole
Synonyms: 6-methyl-1,3-benzoxazole
Molecular Weight (g/mol): 133.15
Molecular Formula: C8H7NO
InChi Key: SZWNDAUMBWLYOQ-UHFFFAOYSA-N
SMILES: CC1=CC=C2N=COC2=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products