accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10541-69-2

CAS RN 10541-69-2

IUPAC Name: 1-(2,5-dichlorophenyl)methanamine
Synonyms: 1-(2,5-dichlorophenyl)methanamine
Molecular Weight (g/mol): 176.04
Molecular Formula: C7H7Cl2N
InChi Key: AKGJLIXNRPNPCH-UHFFFAOYSA-N
SMILES: NCC1=CC(Cl)=CC=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products