CAS RN 10553-31-8
CAS RN 10553-31-8
IUPAC Name:
barium(2+) dibromide
Synonyms:
barium bromide
Molecular Weight (g/mol):
297.14
Molecular Formula:
BaBr2
InChi Key:
NKQIMNKPSDEDMO-UHFFFAOYSA-L
SMILES:
[Br-].[Br-].[Ba++]
1
–
3
of
3
results
1
Barium bromide, 99%, pure, anhydrous, Thermo Scientific Chemicals
CAS: 10553-31-8 Molecular Formula: BaBr2 Molecular Weight (g/mol): 297.16
2
Barium bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
CAS: 10553-31-8 MDL Number: MFCD00003444
3
Barium bromide, anhydrous, 99%, Thermo Scientific Chemicals
CAS: 10553-31-8 MDL Number: MFCD00003444