accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10597-60-1

CAS RN 10597-60-1

IUPAC Name: 4-(2-hydroxyethyl)benzene-1,2-diol
Synonyms: hydroxytyrosol dopet
Molecular Weight (g/mol): 154.17
Molecular Formula: C8H10O3
InChi Key: JUUBCHWRXWPFFH-UHFFFAOYSA-N
SMILES: OCCC1=CC=C(O)C(O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products