man

Test

CAS RN 105973-51-1

OHNH

CAS RN 105973-51-1

IUPAC Name: (3R)-1-benzyl-3-hydroxypiperidin-1-ium
Synonyms: (3R)-1-benzyl-3-hydroxypiperidin-1-ium
Molecular Weight (g/mol): 192.28
Molecular Formula: C12H18NO
InChi Key: UTTCOAGPVHRUFO-GFCCVEGCSA-O
SMILES: O[C@@H]1CCC[NH+](CC2=CC=CC=C2)C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results