man

Test

CAS RN 105994-75-0

CH3NNOOH

CAS RN 105994-75-0

IUPAC Name: 1-methyl-5-phenyl-1H-pyrazole-4-carboxylic acid
Synonyms: 1-methyl-5-phenylpyrazole-4-carboxylic acid
Molecular Weight (g/mol): 202.21
Molecular Formula: C11H10N2O2
InChi Key: MXGOKNNGBUOAGF-UHFFFAOYSA-N
SMILES: CN1N=CC(C(O)=O)=C1C1=CC=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results