man

Test

CAS RN 106-44-5

CH3OH

CAS RN 106-44-5

IUPAC Name: 4-methylphenol
Synonyms: P-cresol aluminum salt 4-cresol p-kresol p-toluol p-cresol for synthesis paramethyl phenol ccris 647 potassium salt 4-cresol mixed cresols sodium salt 4-cresol
Molecular Weight (g/mol): 108.14
Molecular Formula: C7H8O
InChi Key: IWDCLRJOBJJRNH-UHFFFAOYSA-N
SMILES: CC1=CC=C(O)C=C1
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
Boiling Point 
  • (4)
  • (6)
  • (4)
Percent Purity 
  • (2)
  • (4)
  • (4)
  • (4)
Grade 
  • (4)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (5)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (4)
  • (6)
  • (4)
  • (2)
  • (4)
  • (4)
  • (4)
  • (4)
  • (4)
  • (1)