man

Test

CAS RN 106-45-6

CH3SH

CAS RN 106-45-6

IUPAC Name: 4-methylbenzene-1-thiol
Synonyms: 4-methylbenzenethiol
Molecular Weight (g/mol): 124.2
Molecular Formula: C7H8S
InChi Key: WLHCBQAPPJAULW-UHFFFAOYSA-N
SMILES: CC1=CC=C(S)C=C1
Boiling Point 
  • (3)
  • (4)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (4)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)