accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 106276-04-4

CAS RN 106276-04-4

IUPAC Name: {2-[2-(morpholin-4-yl)ethoxy]phenyl}methanol
Synonyms: {2-[2-(morpholin-4-yl)ethoxy]phenyl}methanol
Molecular Weight (g/mol): 237.3
Molecular Formula: C13H19NO3
InChi Key: MJDMCSJTOOAYPS-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1OCCN1CCOCC1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products