man

Test

CAS RN 1067-52-3

H3COHSnHH3CCH3CH3

CAS RN 1067-52-3

IUPAC Name: 1,1,1-tributyl-O-methylstannanol
Synonyms: 1,1,1-tributyl-O-methylstannanol
Molecular Weight (g/mol): 323.11
Molecular Formula: C13H32OSn
InChi Key: SIIBSBQSPWMULI-UHFFFAOYSA-N
SMILES: CO.CCCC[SnH](CCCC)CCCC
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)