accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 107-03-9

CAS RN 107-03-9

IUPAC Name: propane-1-thiol
Synonyms: propanethiol n-thiopropyl alcohol sodium salt propanethiol thiopropyl alcohol
Molecular Weight (g/mol): 76.16
Molecular Formula: C3H8S
InChi Key: SUVIGLJNEAMWEG-UHFFFAOYSA-N
SMILES: CCCS
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products