accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 107-05-1

CAS RN 107-05-1

IUPAC Name: 3-chloroprop-1-ene
Synonyms: allyl chloride
Molecular Weight (g/mol): 76.52
Molecular Formula: C3H5Cl
InChi Key: OSDWBNJEKMUWAV-UHFFFAOYSA-N
SMILES: ClCC=C
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (3)

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products