accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 107-37-9

CAS RN 107-37-9

IUPAC Name: trichloro(prop-2-en-1-yl)silane
Synonyms: allytrichlorosilane
Molecular Weight (g/mol): 175.51
Molecular Formula: C3H5Cl3Si
InChi Key: HKFSBKQQYCMCKO-UHFFFAOYSA-N
SMILES: Cl[Si](Cl)(Cl)CC=C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products