man

Test

CAS RN 107-41-5

CH3HOCH3CH3HO

CAS RN 107-41-5

IUPAC Name: 2-methylpentane-2,4-diol
Synonyms: 2-methyl-2,4-pentanediol hexylene glycol, titanium(4+) salt caswell no. 574 titanium(4+) salt hexylene glycol epa pesticide chemical code 068601 MPD hexylene glycol (substance) hexylene glycol pinakon diolane
Molecular Weight (g/mol): 118.18
Molecular Formula: C6H14O2
InChi Key: SVTBMSDMJJWYQN-UHFFFAOYNA-N
SMILES: CC(O)CC(C)(C)O
Molecular Weight (g/mol) 
  • (5)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (4)
  • (4)
Percent Purity 
  • (3)
  • (3)
  • (1)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (3)
  • (5)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)