man

Test

CAS RN 107-95-9

H2NOHO

CAS RN 107-95-9

IUPAC Name: 3-aminopropanoic acid
Synonyms: β alanine monosodium salt β alanine abufene monosodium salt β-alanine 3-aminopropionsaeure H-β-ala-OH β-aminopropionsaeure bAla monopotassium salt β-alanine ω-aminopropionic acid
Molecular Weight (g/mol): 89.09
Molecular Formula: C3H7NO2
InChi Key: UCMIRNVEIXFBKS-UHFFFAOYSA-N
SMILES: NCCC(O)=O
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)