man

Test

CAS RN 1070-83-3

H3CH3CCH3OHO

CAS RN 1070-83-3

IUPAC Name: 3,3-dimethylbutanoic acid
Synonyms: 3,3-dimethylbutanoic acid des-amino t-butyl glycine des-amino T-butyl glycine
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: MLMQPDHYNJCQAO-UHFFFAOYSA-N
SMILES: CC(C)(C)CC(O)=O
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)