accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1071-23-4

CAS RN 1071-23-4

IUPAC Name: (2-aminoethoxy)phosphonic acid
Synonyms: O-phosphoethanolamine colaminphosphoric acid phosphorylethanolamine magnesium (1:1) salt EAP OPE PEA phosphorylethanolamine phosphoryethanolamine ethanolamine acid phosphate ethanolamine O-phosphate
Molecular Weight (g/mol): 141.06
Molecular Formula: C2H8NO4P
InChi Key: SUHOOTKUPISOBE-UHFFFAOYSA-N
SMILES: NCCOP(O)(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products