accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1071-27-8

CAS RN 1071-27-8

IUPAC Name: 4-(trichlorosilyl)butanenitrile
Synonyms: 4-(trichlorosilyl)butanenitrile
Molecular Weight (g/mol): 202.53
Molecular Formula: C4H6Cl3NSi
InChi Key: HMFFOEBLYHLRQN-UHFFFAOYSA-N
SMILES: Cl[Si](Cl)(Cl)CCCC#N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products