man

Test

CAS RN 1071-46-1

H3COOOHO

CAS RN 1071-46-1

IUPAC Name: 3-ethoxy-3-oxopropanoic acid
Synonyms: ethyl hydrogen malonate
Molecular Weight (g/mol): 132.12
Molecular Formula: C5H8O4
InChi Key: HGINADPHJQTSKN-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(O)=O
Boiling Point 
  • (7)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (7)
Percent Purity 
  • (4)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (7)
  • (3)
  • (3)
  • (7)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)