man

Test

CAS RN 1072-71-5

SNHNHSS

CAS RN 1072-71-5

IUPAC Name: 1,3,4-thiadiazolidine-2,5-dithione
Synonyms: 1,3,4-thiadiazolidine-2,5-dithione
Molecular Weight (g/mol): 150.23
Molecular Formula: C2H2N2S3
InChi Key: BIGYLAKFCGVRAN-UHFFFAOYSA-N
SMILES: S=C1NNC(=S)S1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results