CAS RN 1072-72-6
CAS RN 1072-72-6
IUPAC Name:
thian-4-one
Synonyms:
thian-4-one
Molecular Weight (g/mol):
116.18
Molecular Formula:
C5H8OS
InChi Key:
OVRJVKCZJCNSOW-UHFFFAOYSA-N
SMILES:
O=C1CCSCC1
1
–
2
of
2
results
1
Tetrahydro-4H-thiopyran-4-one, 99%, Thermo Scientific Chemicals
CAS: 1072-72-6 Molecular Formula: C5H8OS Molecular Weight (g/mol): 116.18 MDL Number: MFCD00006660 InChI Key: OVRJVKCZJCNSOW-UHFFFAOYSA-N Synonym: tetrahydrothiopyran-4-one,tetrahydro-4h-thiopyran-4-one,4-oxothiane,4-thiacyclohexanone,penthianone,4h-thiopyran-4-one, tetrahydro,1-thiacyclohexan-4-one,dihydro-2h-thiopyran-4 3h-one,4-oxotetrahydrothiopyran,tetrahydro-4-thiopyrone PubChem CID: 66173 IUPAC Name: thian-4-one SMILES: O=C1CCSCC1