accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1072-82-8

CAS RN 1072-82-8

IUPAC Name: 1-(1H-pyrrol-3-yl)ethan-1-one
Synonyms: 1-(1H-pyrrol-3-yl)ethanone
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: KHHSXHXUQVNBGA-UHFFFAOYSA-N
SMILES: CC(=O)C1=CNC=C1
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products