man

Test

CAS RN 1072952-34-1

BNCH3HOOHHCl

CAS RN 1072952-34-1

IUPAC Name: (2-methylpyridin-3-yl)boronic acid hydrochloride
Synonyms: 2-methylpyridin-3-ylboronic acid hydrochloride
Molecular Weight (g/mol): 173.4
Molecular Formula: C6H9BClNO2
InChi Key: OAMCCRLFDPJOPN-UHFFFAOYSA-N
SMILES: Cl.CC1=C(C=CC=N1)B(O)O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results