accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1073-23-0

CAS RN 1073-23-0

IUPAC Name: 2,6-dimethylpyridin-1-ium-1-olate
Synonyms: 2,6-dimethylpyridin-1-ium-1-olate
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: LIDGFHXPUOJZMK-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=[N+]1[O-]
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products