man

Test

CAS RN 1073-26-3

CH3ONH

CAS RN 1073-26-3

IUPAC Name: 1-(1H-pyrrol-2-yl)propan-1-one
Synonyms: 1-propanone, 1-pyrrol-2-yl-
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: AEJPPSRYXGEVDT-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=CN1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results