man

Test

CAS RN 1073-29-6

H3CSHO

CAS RN 1073-29-6

IUPAC Name: 2-(methylsulfanyl)phenol
Synonyms: 2-(methylsulfanyl)phenol
Molecular Weight (g/mol): 140.2
Molecular Formula: C7H8OS
InChi Key: SOOARYARZPXNAL-UHFFFAOYSA-N
SMILES: CSC1=CC=CC=C1O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results