man

Test

CAS RN 1073353-73-7

BOOH3CCH3H3CH3CNOCH3Cl

CAS RN 1073353-73-7

IUPAC Name: 3-chloro-2-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 3-chloro-2-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 269.53
Molecular Formula: C12H17BClNO3
InChi Key: JYVZFQGNBUYGIH-UHFFFAOYSA-N
SMILES: COC1=NC=CC(B2OC(C)(C)C(C)(C)O2)=C1Cl

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results