man

Test

CAS RN 1075-06-5

OHHOO

CAS RN 1075-06-5

IUPAC Name: 2,2-dihydroxy-1-phenylethan-1-one
Synonyms: phenylglyoxal
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: NBIBDIKAOBCFJN-UHFFFAOYSA-N
SMILES: OC(O)C(=O)C1=CC=CC=C1
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)