accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 107575-60-0

CAS RN 107575-60-0

IUPAC Name: ethyl 6-amino-5-methoxy-1H-indole-2-carboxylate
Synonyms: ethyl 6-amino-5-methoxy-1H-indole-2-carboxylate
Molecular Weight (g/mol): 234.26
Molecular Formula: C12H14N2O3
InChi Key: DIEUGINJYISDCB-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=CC(OC)=C(N)C=C2N1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products