man

Test

CAS RN 107703-78-6

HON

CAS RN 107703-78-6

IUPAC Name: phenyl[1-(2-phenylethyl)piperidin-4-yl]methanol
Synonyms: glemanserin 格来色林
Molecular Weight (g/mol): 295.43
Molecular Formula: C20H25NO
InChi Key: AXNGJCOYCMDPQG-UHFFFAOYNA-N
SMILES: OC(C1CCN(CCC2=CC=CC=C2)CC1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results