accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 107819-90-9

CAS RN 107819-90-9

IUPAC Name: tert-butyl N-({[(tert-butoxy)carbonyl]amino}(methylsulfanyl)methylidene)carbamate
Synonyms: tert-butyl N-{[(tert-butoxycarbonyl)amino](methylsulfanyl)methylidene}carbamate
Molecular Weight (g/mol): 290.38
Molecular Formula: C12H22N2O4S
InChi Key: UQJXXWHAJKRDKY-UHFFFAOYSA-N
SMILES: CSC(NC(=O)OC(C)(C)C)=NC(=O)OC(C)(C)C

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products