accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 108-05-4

CAS RN 108-05-4

IUPAC Name: ethenyl acetate
Synonyms: vinyl acetate acetate de vinyle zeset T vinylacetat vinyl A monomer vinylazetat vinile (acetato di) vinyl acetate monomer vinyl acetate H.Q. essigsaeurevinylester
Molecular Weight (g/mol): 86.09
Molecular Formula: C4H6O2
InChi Key: XTXRWKRVRITETP-UHFFFAOYSA-N
SMILES: CC(=O)OC=C
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (7)
Percent Purity 
  • (3)
  • (4)

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (7)
  • (3)
  • (4)
  • (5)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products