man

Test

CAS RN 108-16-7

CH3HONCH3H3C

CAS RN 108-16-7

IUPAC Name: 1-(dimethylamino)propan-2-ol
Synonyms: dimepranol dimethyl isopropanolamine
Molecular Weight (g/mol): 103.17
Molecular Formula: C5H13NO
InChi Key: NCXUNZWLEYGQAH-UHFFFAOYNA-N
SMILES: CC(O)CN(C)C
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)