CAS RN 1080028-74-5
CAS RN 1080028-74-5
IUPAC Name:
tert-butyl 4-[(4-formylphenyl)methoxy]piperidine-1-carboxylate
Synonyms:
tert-butyl 4-[(4-formylphenyl)methoxy]piperidine-1-carboxylate
Molecular Weight (g/mol):
319.4
Molecular Formula:
C18H25NO4
InChi Key:
WQFHJESQGSVMDW-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)N1CCC(CC1)OCC1=CC=C(C=O)C=C1
1
–
1
of
1
results
1
tert-butyl 4-(4-formylbenzyloxy)piperidine-1-carboxylate, 90%, Thermo Scientific™
CAS: 1080028-74-5 Molecular Formula: C18H25NO4 Molecular Weight (g/mol): 319.401 MDL Number: MFCD11841070 InChI Key: WQFHJESQGSVMDW-UHFFFAOYSA-N Synonym: tert-butyl 4-4-formylbenzyloxy piperidine-1-carboxylate,tert-butyl 4-4-formylphenyl methoxy piperidine-1-carboxylate,1-4-formylbenzyl oxy piperidine, n-boc protected,tert-butyl 4-4-formylbenzyl oxy piperidine-1-carboxylate,4-1-tert-butoxycarbonyl piperidin-4-yl oxy methyl benzaldehyde PubChem CID: 33589526 IUPAC Name: tert-butyl 4-[(4-formylphenyl)methoxy]piperidine-1-carboxylate SMILES: CC(C)(C)OC(=O)N1CCC(CC1)OCC2=CC=C(C=C2)C=O