accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 108238-09-1

CAS RN 108238-09-1

IUPAC Name: (2-phenoxyphenyl)boronic acid
Synonyms: 2-phenoxyphenylboronic acid
Molecular Weight (g/mol): 214.03
Molecular Formula: C12H11BO3
InChi Key: AVOWPOFIQZSVGV-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC=C1OC1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products