CAS RN 1085-12-7
CAS RN 1085-12-7
IUPAC Name:
heptyl 4-hydroxybenzoate
Synonyms:
heptylparaben nipaheptyl p-oxybenzoesaureheptylester staypro WS 7 heptyl P-hydroxybenzoate
Molecular Weight (g/mol):
236.31
Molecular Formula:
C14H20O3
InChi Key:
ZTJORNVITHUQJA-UHFFFAOYSA-N
SMILES:
CCCCCCCOC(=O)C1=CC=C(O)C=C1
1
–
1
of
1
results
1
n-Heptyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals
CAS: 1085-12-7 Molecular Formula: C14H20O3 Molecular Weight (g/mol): 236.31 MDL Number: MFCD00016481 InChI Key: ZTJORNVITHUQJA-UHFFFAOYSA-N Synonym: heptylparaben,heptyl paraben,heptyl p-hydroxybenzoate,n-heptyl 4-hydroxybenzoate,nipaheptyl,staypro ws 7,n-heptyl p-hydroxybenzoate,benzoic acid, 4-hydroxy-, heptyl ester,p-hydroxybenzoic acid heptyl ester,p-oxybenzoesaureheptylester PubChem CID: 14138 ChEBI: CHEBI:34788 IUPAC Name: heptyl 4-hydroxybenzoate SMILES: CCCCCCCOC(=O)C1=CC=C(O)C=C1