CAS RN 108963-96-8
CAS RN 108963-96-8
IUPAC Name:
1-tert-butyl 2-methyl 5-oxopyrrolidine-1,2-dicarboxylate
Synonyms:
1-tert-butyl 2-methyl 5-oxopyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol):
243.26
Molecular Formula:
C11H17NO5
InChi Key:
FNTAOUUEQHKLIU-UHFFFAOYNA-N
SMILES:
COC(=O)C1CCC(=O)N1C(=O)OC(C)(C)C
1
–
1
of
1
results
1
Methyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate, 98%, Thermo Scientific Chemicals
CAS: 108963-96-8 Molecular Formula: C11H17NO5 Molecular Weight (g/mol): 243.259 MDL Number: MFCD06809720 InChI Key: FNTAOUUEQHKLIU-ZETCQYMHSA-N Synonym: boc-l-pyroglutamic acid methyl ester,boc-pyr-ome,s-1-tert-butyl 2-methyl 5-oxopyrrolidine-1,2-dicarboxylate,methyl l-n-boc-pyroglutamate,1-tert-butyl 2-methyl 2s-5-oxopyrrolidine-1,2-dicarboxylate,methyl boc-l-pyroglutamate,s-5-oxo-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester 2-methyl ester,1-tert-butyl 2-methyl s-5-oxopyrrolidine-1,2-dicarboxylate,boc-pglu-ome,pubchem12135 PubChem CID: 10868485 IUPAC Name: 1-O-tert-butyl 2-O-methyl (2S)-5-oxopyrrolidine-1,2-dicarboxylate SMILES: CC(C)(C)OC(=O)N1C(CCC1=O)C(=O)OC