man

Test

CAS RN 109-80-8

SHHS

CAS RN 109-80-8

IUPAC Name: propane-1,3-dithiol
Synonyms: 1,3-propanedithiol trimethylenedithioglycol 1,3-propanedimercaptan dithiotrimethyleneglycol trimethylene dimercaptan
Molecular Weight (g/mol): 108.22
Molecular Formula: C3H8S2
InChi Key: ZJLMKPKYJBQJNH-UHFFFAOYSA-N
SMILES: SCCCS
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)