man

Test

CAS RN 109431-87-0

CH3H3CCH3OONHO

CAS RN 109431-87-0

IUPAC Name: tert-butyl 3-hydroxypyrrolidine-1-carboxylate
Synonyms: tert-butyl 3-hydroxypyrrolidine-1-carboxylate
Molecular Weight (g/mol): 187.24
Molecular Formula: C9H17NO3
InChi Key: APCBTRDHCDOPNY-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCC(O)C1
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results