CAS RN 110-75-8
CAS RN 110-75-8
IUPAC Name:
(2-chloroethoxy)ethene
Synonyms:
2-chloroethyl vinyl ether
Molecular Weight (g/mol):
106.55
Molecular Formula:
C4H7ClO
InChi Key:
DNJRKFKAFWSXSE-UHFFFAOYSA-N
SMILES:
ClCCOC=C