man

Test

CAS RN 110-88-3

OOO

CAS RN 110-88-3

IUPAC Name: 1,3,5-trioxane
Synonyms: trioxane marvosan formagene triossimetilene s-trixane trioxymethylen trimer formaldehyde formaldehyde, trimer metaformaldehyde triformol
Molecular Weight (g/mol): 90.08
Molecular Formula: C3H6O3
InChi Key: BGJSXRVXTHVRSN-UHFFFAOYSA-N
SMILES: C1OCOCO1
Boiling Point 
  • (5)
Physical Form 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)