man

Test

CAS RN 1100-88-5

ClP

CAS RN 1100-88-5

IUPAC Name: benzyltriphenylphosphanium chloride
Synonyms: benzyltriphenylphosphanium chloride
Molecular Weight (g/mol): 388.87
Molecular Formula: C25H22ClP
InChi Key: USFRYJRPHFMVBZ-UHFFFAOYSA-M
SMILES: [Cl-].C(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)