CAS RN 111-13-7
CAS RN 111-13-7
IUPAC Name:
octan-2-one
Synonyms:
2-octanone
Molecular Weight (g/mol):
128.22
Molecular Formula:
C8H16O
InChi Key:
ZPVFWPFBNIEHGJ-UHFFFAOYSA-N
SMILES:
CCCCCCC(C)=O
1
–
2
of
2
results
1
2-Octanone, 99+%, Thermo Scientific Chemicals
CAS: 111-13-7 MDL Number: MFCD00009540 InChI Key: ZPVFWPFBNIEHGJ-UHFFFAOYSA-N Synonym: 2-octanone,n-hexyl methyl ketone,hexyl methyl ketone,methyl hexyl ketone,methyl n-hexyl ketone,octanone,2-oxooctane,2-octanone natural,fema number 2802,unii-j2g84h29af PubChem CID: 8093 ChEBI: CHEBI:87434 IUPAC Name: octan-2-one SMILES: CCCCCCC(=O)C
Promotions are available
2
2-Octanone, 98%, Thermo Scientific Chemicals
CAS: 111-13-7 Molecular Formula: C8H16O Molecular Weight (g/mol): 128.215 MDL Number: MFCD00009540 InChI Key: ZPVFWPFBNIEHGJ-UHFFFAOYSA-N Synonym: 2-octanone,n-hexyl methyl ketone,hexyl methyl ketone,methyl hexyl ketone,methyl n-hexyl ketone,octanone,2-oxooctane,2-octanone natural,fema number 2802,unii-j2g84h29af PubChem CID: 8093 ChEBI: CHEBI:87434 IUPAC Name: octan-2-one SMILES: CCCCCCC(=O)C