CAS RN 111-24-0
CAS RN 111-24-0
IUPAC Name:
1,5-dibromopentane
Synonyms:
pentane, 1,5-dibromo-
Molecular Weight (g/mol):
229.94
Molecular Formula:
C5H10Br2
InChi Key:
IBODDUNKEPPBKW-UHFFFAOYSA-N
SMILES:
BrCCCCCBr
1
–
2
of
2
results
1
1,5-Dibromopentane, 97%, Thermo Scientific Chemicals
CAS: 111-24-0 Molecular Formula: C5H10Br2 Molecular Weight (g/mol): 229.94 MDL Number: MFCD00000268 InChI Key: IBODDUNKEPPBKW-UHFFFAOYSA-N Synonym: pentamethylene bromide,pentane, 1,5-dibromo,pentamethylene dibromide,unii-b8q228qys1,pentane-1,5-dibromide,1,5-dibromo-pentane,1,5-pentane dibromide,pentane,1,5-dibromo,br ch2 5br,ccris 8918 PubChem CID: 8100 IUPAC Name: 1,5-dibromopentane SMILES: C(CCBr)CCBr
2
1,5-Dibromopentane, 98%, Thermo Scientific Chemicals
CAS: 111-24-0 Molecular Formula: C5H10Br2 Molecular Weight (g/mol): 229.943 MDL Number: MFCD00000268 InChI Key: IBODDUNKEPPBKW-UHFFFAOYSA-N PubChem CID: 8100 IUPAC Name: 1,5-dibromopentane SMILES: C(CCBr)CCBr