man

Test

CAS RN 111-62-6

H3COOCH3

CAS RN 111-62-6

IUPAC Name: ethyl (9E)-octadec-9-enoate
Synonyms: ethyl oleate
Molecular Weight (g/mol): 310.52
Molecular Formula: C20H38O2
InChi Key: LVGKNOAMLMIIKO-VAWYXSNFSA-N
SMILES: CCCCCCCC\C=C\CCCCCCCC(=O)OCC
Boiling Point 
  • (3)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)