CAS RN 111247-62-2
CAS RN 111247-62-2
IUPAC Name:
2-[4-(chloromethyl)piperidin-1-yl]pyrimidine
Synonyms:
2-[4-(chloromethyl)piperidin-1-yl]pyrimidine
Molecular Weight (g/mol):
211.69
Molecular Formula:
C10H14ClN3
InChi Key:
XEOONRMTEFSPSW-UHFFFAOYSA-N
SMILES:
ClCC1CCN(CC1)C1=NC=CC=N1
1
–
1
of
1
results
1
2-[4-(chloromethyl)piperidino]pyrimidine, Thermo Scientific™
CAS: 111247-62-2 Molecular Formula: C10H14ClN3 Molecular Weight (g/mol): 211.693 InChI Key: XEOONRMTEFSPSW-UHFFFAOYSA-N Synonym: 2-4-chloromethyl piperidin-1-yl pyrimidine,2-4-chloromethyl piperidino pyrimidine,2-4-chloromethyl-piperidin-1-yl-pyrimidine,2-4-chloromethyl piperidyl pyrimidine,2-4-chloromethylpiperidin-1-yl pyrimidine PubChem CID: 14554091 IUPAC Name: 2-[4-(chloromethyl)piperidin-1-yl]pyrimidine SMILES: C1CN(CCC1CCl)C2=NC=CC=N2