CAS RN 1115-69-1
CAS RN 1115-69-1
IUPAC Name:
2-acetamidopropanoic acid
Synonyms:
2-acetamidopropanoic acid
Molecular Weight (g/mol):
131.13
Molecular Formula:
C5H9NO3
InChi Key:
KTHDTJVBEPMMGL-UHFFFAOYNA-N
SMILES:
CC(NC(C)=O)C(O)=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals N-Acetyl-DL-alanine, 97+%
CAS: 1115-69-1 Molecular Formula: C5H9NO3 Molecular Weight (g/mol): 131.131 MDL Number: MFCD00037238 InChI Key: KTHDTJVBEPMMGL-UHFFFAOYSA-N Synonym: n-acetyl-dl-alanine,ac-dl-ala-oh,2-acetylaminopropionic acid,n-acetylalanine,acetyl-dl-alanine,dl-alanine, n-acetyl,2-acetylamino-propionic acid,n-acetyl-s-alanine,n-alpha-acetyl-dl-alanine,n-acetyl-dl-2-aminopropionic acid PubChem CID: 7345 IUPAC Name: 2-acetamidopropanoic acid SMILES: CC(C(=O)O)NC(=O)C